siRNA Details
| Gene Symbol: 9330133O14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK033985.1 | - | 1 | 1485 | ||
| AK049744.1 | - | 1 | 1543 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | NM_176948.1 | - | 1543 | Chr. - - [-] on Build | ||

9330133O14Rik
Genomic Map