siRNA Details
| Gene Symbol: 6430573F11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176952.4 | NP_795926.2 | 4 | 1482 | ||
| GenBank mRNA | AK032504.1 | - | 7 | 2288 | ||
| AK141601.1 | - | 1856 | ||||
| BC052184.1 | - | 4 | 1482 | 6336362 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

6430573F11Rik| Gene Symbol: 6430573F11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176952.4 | NP_795926.2 | 4 | 1482 | ||
| GenBank mRNA | AK032504.1 | - | 7 | 2288 | ||
| AK141601.1 | - | 1856 | ||||
| BC052184.1 | - | 4 | 1482 | 6336362 | ||