siRNA Details
| Gene Symbol: 9330132A10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK033982.1 | - | 2302 | |||
| AK083197.1 | - | 2076 | ||||
| BC098197.1 | - | 2029 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9330132A10Rik| Gene Symbol: 9330132A10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK033982.1 | - | 2302 | |||
| AK083197.1 | - | 2076 | ||||
| BC098197.1 | - | 2029 | ||||