siRNA Details
| Gene Symbol: Rab9b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176971.2 | NP_795945.1 | 3 | 1214 | ||
| GenBank mRNA | AK034442.1 | - | 3 | 1215 | ||
| AK049693.1 | - | 3 | 1241 | |||
| AK161867.1 | - | 3 | 1436 | |||
| BC107383.2 | - | 3 | 1186 | 40057619 | ||

Rab9b
Genomic Map