siRNA Details
| Gene Symbol: Rab9b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176971.2 | NP_795945.1 | 3 | 2542 | ||
| GenBank mRNA | AK034442.1 | - | 3 | 2543 | ||
| AK049693.1 | - | 3 | 2569 | |||
| AK161867.1 | - | 3 | 2764 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rab9b| Gene Symbol: Rab9b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176971.2 | NP_795945.1 | 3 | 2542 | ||
| GenBank mRNA | AK034442.1 | - | 3 | 2543 | ||
| AK049693.1 | - | 3 | 2569 | |||
| AK161867.1 | - | 3 | 2764 | |||