siRNA Details
| Gene Symbol: 5930422O12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK031179.1 | - | 1 | 2649 | ||
| BB037066.2 | - | 1 | 82 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

5930422O12Rik| Gene Symbol: 5930422O12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK031179.1 | - | 1 | 2649 | ||
| BB037066.2 | - | 1 | 82 | |||