siRNA Details
| Gene Symbol: Topbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176979.5 | NP_795953.2 | 3 | 243 | ||
| GenBank mRNA | AK084654.1 | - | 3 | 346 | ||
| AK085031.1 | - | 3 | 382 | |||
| AK129104.1 | - | 3 | 355 | |||
| BC049797.1 | - | 3 | 243 | 6510112 | ||

Topbp1
Genomic Map