siRNA Details
| Gene Symbol: AIM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004833.1 | NP_004824.1 | 2, 3 | 508 | ||
| XM_017002848.1 | XP_016858337.1 | 3, 4 | 1735 | |||
| GenBank mRNA | AF024714.1 | - | 2, 3 | 508 | ||
| AK292042.1 | - | 2, 3 | 417 | |||
| BC010940.1 | - | 2, 3 | 510 | 4080360 | ||

AIM2
Genomic Map