siRNA Details
| Gene Symbol: Tigar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177003.5 | NP_795977.1 | 3 | 263 | ||
| GenBank mRNA | AK036082.1 | - | 3 | 254 | ||
| AK078900.1 | - | 3 | 254 | |||
| AK145896.1 | - | 3 | 248 | |||
| AK163290.1 | - | 3 | 239 | |||
| AK165614.1 | - | 3 | 264 | |||
| BC147705.1 | - | 3 | 253 | 9087464 | ||
| BC147718.1 | - | 3 | 253 | 9087477 | ||

Tigar
Genomic Map