siRNA Details
| Gene Symbol: Smyd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001102611.1 | NP_001096081.1 | 5 | 1839 | ||
| GenBank mRNA | AK030380.1 | - | 5 | 1676 | ||
| AK089959.1 | - | 5 | 1775 | |||
| AK154507.1 | - | 4 | 1525 | |||
| AK173306.1 | - | 3 | 1261 | |||
| BC049810.1 | - | 5 | 1690 | |||
| BC095952.1 | - | 5 | 1736 | 5037842 | ||
| BC130220.1 | - | 2 | 1176 | 40141368 | ||

Smyd4
Genomic Map