siRNA Details
| Gene Symbol: Tmem229a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177013.3 | NP_795987.2 | 1 | 2286 | ||
| GenBank mRNA | AK032121.1 | - | 1 | 296 | ||
| AK087465.1 | - | 1 | 2254 | |||
| AK140056.1 | - | 1 | 2237 | |||
| AK161777.1 | - | 1 | 2460 | |||

Tmem229a
Genomic Map