siRNA Details
| Gene Symbol: E130114P18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK053617.1 | - | 1044 | |||
| AK082601.1 | - | 3 | 886 | |||
| AK086199.1 | - | 1220 | ||||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | NM_177023.1 | - | 1220 | Chr. - - [-] on Build | ||

E130114P18Rik
Genomic Map