siRNA Details
| Gene Symbol: Cobll1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027225.1 | NP_081501.1 | 3, 4 | 649 | ||
| NM_177025.5 | NP_795999.2 | 3, 4 | 649 | |||
| GenBank mRNA | AK037010.1 | - | 4, 5 | 640 | ||
| AK052533.1 | - | 3, 4 | 598 | |||
| AK144943.1 | - | 4, 5 | 693 | |||
| AK164624.1 | - | 4, 5 | 579 | |||
| AK173076.1 | - | 3, 4 | 480 | |||
| AY308746.1 | - | 3, 4 | 649 | |||
| BC067007.1 | - | 3, 4 | 646 | 5702930 | ||

Cobll1
Genomic Map