siRNA Details
| Gene Symbol: Cobll1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027225.1 | NP_081501.1 | 5 | 971 | ||
| NM_177025.5 | NP_795999.2 | 5 | 971 | |||
| GenBank mRNA | AK037010.1 | - | 6 | 962 | ||
| AK052533.1 | - | 5 | 920 | |||
| AK144943.1 | - | 6 | 1015 | |||
| AK164624.1 | - | 6 | 901 | |||
| AK173076.1 | - | 5 | 802 | |||
| AY308746.1 | - | 5 | 971 | |||
| BC067007.1 | - | 5 | 968 | 5702930 | ||

Cobll1
Genomic Map