siRNA Details
| Gene Symbol: Kif6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177052.3 | NP_796026.2 | 7 | 728 | ||
| GenBank mRNA | AB001434.1 | - | 4 | 387 | ||
| AK043404.1 | - | 7 | 685 | |||
| AK083760.1 | - | 7 | 765 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kif6| Gene Symbol: Kif6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177052.3 | NP_796026.2 | 7 | 728 | ||
| GenBank mRNA | AB001434.1 | - | 4 | 387 | ||
| AK043404.1 | - | 7 | 685 | |||
| AK083760.1 | - | 7 | 765 | |||