siRNA Details
| Gene Symbol: Rnf222 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177060.3 | NP_796034.2 | 3 | 1041 | ||
| GenBank mRNA | AK028294.1 | - | 3 | 1042 | ||
| AK037019.1 | - | 1 | 773 | |||
| AK037028.1 | - | 2 | 813 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnf222| Gene Symbol: Rnf222 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177060.3 | NP_796034.2 | 3 | 1041 | ||
| GenBank mRNA | AK028294.1 | - | 3 | 1042 | ||
| AK037019.1 | - | 1 | 773 | |||
| AK037028.1 | - | 2 | 813 | |||