siRNA Details
| Gene Symbol: BC106179 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK041922.1 | - | 3 | 731 | ||
| AK161755.1 | - | 2 | 1305 | |||
| BC106179.1 | - | 2 | 1301 | 6509459 | ||
 Back To Top
 Back To TopPlease enter the information below and press OK to send your cart to Core Services for purchase.

 BC106179
BC106179| Gene Symbol: BC106179 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK041922.1 | - | 3 | 731 | ||
| AK161755.1 | - | 2 | 1305 | |||
| BC106179.1 | - | 2 | 1301 | 6509459 | ||
 Back To Top
 Back To Top