siRNA Details
| Gene Symbol: 6030445D17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK031509.1 | - | 1 | 1433 | ||
| AK033467.1 | - | 1 | 1346 | |||
| BB039760.2 | - | 1 | 378 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

6030445D17Rik| Gene Symbol: 6030445D17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK031509.1 | - | 1 | 1433 | ||
| AK033467.1 | - | 1 | 1346 | |||
| BB039760.2 | - | 1 | 378 | |||