siRNA Details
| Gene Symbol: Hykk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177351.4 | NP_796325.2 | 5 | 2580 | ||
| GenBank mRNA | AK085527.1 | - | 5 | 2590 | ||
| AK138851.1 | - | 5 | 2566 | |||
| AK162425.1 | - | 5 | 2581 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hykk| Gene Symbol: Hykk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177351.4 | NP_796325.2 | 5 | 2580 | ||
| GenBank mRNA | AK085527.1 | - | 5 | 2590 | ||
| AK138851.1 | - | 5 | 2566 | |||
| AK162425.1 | - | 5 | 2581 | |||