siRNA Details
| Gene Symbol: Zfp945 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110254.1 | NP_001103724.1 | 5 | 4342 | ||
| NM_177358.4 | NP_796332.2 | 6 | 4463 | |||
| GenBank mRNA | AK034838.1 | - | 1 | 2854 | ||
| AK041739.1 | - | 1 | 1492 | |||
| AK044245.1 | - | 1 | 1262 | |||
| AK164594.1 | - | 1 | 1069 | |||

Zfp945
Genomic Map