siRNA Details
| Gene Symbol: Zfp799 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293229.1 | NP_001280158.1 | 2, 3 | 295 | ||
| NM_177359.5 | NP_796333.3 | 2, 3 | 295 | |||
| GenBank mRNA | AK031686.1 | - | 2, 3 | 283 | ||
| AK083640.1 | - | 2, 3 | 272 | |||
| AK137037.1 | - | 2, 3 | 272 | |||
| BC062813.1 | - | 2, 3 | 249 | |||
| BC096552.1 | - | 2, 3 | 245 | 30618827 | ||

Zfp799
Genomic Map