siRNA Details
Gene Symbol: Dmrt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177360.3 | NP_796334.2 | 2 | 2262 | ||
GenBank mRNA | AF541936.1 | - | 2 | 2262 | ||
BC052041.1 | - | 2 | 2256 | 6404988 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dmrt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177360.3 | NP_796334.2 | 2 | 2262 | ||
GenBank mRNA | AF541936.1 | - | 2 | 2262 | ||
BC052041.1 | - | 2 | 2256 | 6404988 |