siRNA Details
| Gene Symbol: Ofd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177429.3 | NP_803178.2 | 16 | 2368 | ||
| GenBank mRNA | AJ438159.1 | - | 16 | 2219 | ||
| BC119070.1 | - | 16 | 2051 | 8733819 | ||
| BC120487.1 | - | 16 | 2051 | 40129410 | ||

Ofd1
Genomic Map