siRNA Details
| Gene Symbol: 4833427G06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177702.3 | NP_808370.1 | 4, 5 | 505 | ||
| GenBank mRNA | AK014780.1 | - | 4, 5 | 394 | ||
| AK132837.1 | - | 4, 5 | 468 | |||
| BC048587.1 | - | 4, 5 | 505 | 6707436 | ||

4833427G06Rik
Genomic Map