siRNA Details
| Gene Symbol: Sytl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290728.1 | NP_001277657.1 | 15 | 1971 | ||
| NM_177704.3 | NP_808372.1 | 16 | 2037 | |||
| GenBank mRNA | AB098160.1 | - | 16 | 2036 | ||
| AB098161.1 | - | 15 | 1970 | |||
| BC150158.1 | - | 16 | 2040 | 40141504 | ||
| BC150480.1 | - | 16 | 2040 | 40141505 | ||

Sytl5
Genomic Map