siRNA Details
| Gene Symbol: Rtn4rl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177708.5 | NP_808376.1 | 2 | 1837 | ||
| GenBank mRNA | AK033286.1 | - | 2 | 1835 | ||
| AK140456.1 | - | 2 | 1838 | |||
| BC030471.1 | - | 2 | 2041 | 5368560 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rtn4rl1| Gene Symbol: Rtn4rl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177708.5 | NP_808376.1 | 2 | 1837 | ||
| GenBank mRNA | AK033286.1 | - | 2 | 1835 | ||
| AK140456.1 | - | 2 | 1838 | |||
| BC030471.1 | - | 2 | 2041 | 5368560 | ||