siRNA Details
| Gene Symbol: POLR1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318876.1 | NP_001305805.1 | 2, 3 | 220 | ||
| NM_203290.3 | NP_976035.1 | 2, 3 | 220 | |||
| XM_011515000.2 | XP_011513302.1 | 2, 3 | 161 | |||
| GenBank mRNA | AF008442.1 | - | 2, 3 | 146 | ||
| AF047441.1 | - | 2, 3 | 156 | |||
| AK293561.1 | - | 2, 3 | 146 | |||
| BC008118.1 | - | 2, 3 | 111 | |||
| BC008863.2 | - | 2, 3 | 146 | 4128104 | ||
| BM828768.1 | - | 2, 3 | 211 | |||
| DW415914.1 | - | 2, 3 | 239 | |||

POLR1C
Genomic Map