siRNA Details
| Gene Symbol: Ssh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291190.1 | NP_001278119.1 | 14 | 3973 | ||
| NM_177710.4 | NP_808378.2 | 15 | 3837 | |||
| GenBank mRNA | AB099288.1 | - | 15 | 3837 | ||
| AK155232.1 | - | 14 | 3964 | |||
| AK173243.1 | - | 4 | 2701 | |||
| BC030940.1 | - | 1 | 385 | |||
| BC141392.1 | - | 15 | 3831 | 9056005 | ||
| BC141393.1 | - | 15 | 3831 | 9056006 | ||

Ssh2
Genomic Map