siRNA Details
| Gene Symbol: Kctd12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177715.4 | NP_808383.3 | 1 | 2987 | ||
| GenBank mRNA | AK032276.1 | - | 1 | 872 | ||
| AK035535.1 | - | 1 | 2988 | |||
| AK049420.1 | - | 1 | 1112 | |||
| AK086988.1 | - | 1 | 859 | |||
| AK172581.1 | - | 1 | 2987 | |||
| AK173260.1 | - | 1 | 2492 | |||
| BC116647.1 | - | 2 | 2374 | |||
| BC116648.1 | - | 2 | 2375 | |||

Kctd12
Genomic Map