siRNA Details
| Gene Symbol: Impad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177730.3 | NP_808398.1 | 1 | 465 | ||
| GenBank mRNA | AK045511.1 | - | 1 | 515 | ||
| BC048776.1 | - | 1 | 465 | 6439769 | ||
| BC138209.1 | - | 1 | 322 | 8861228 | ||
| BC145952.1 | - | 1 | 322 | 40131255 | ||

Impad1
Genomic Map