siRNA Details
| Gene Symbol: Klhl18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177771.5 | NP_808439.2 | 6 | 891 | ||
| GenBank mRNA | AK044774.1 | - | 6 | 1002 | ||
| AK173029.1 | - | 6 | 891 | |||
| AK198584.1 | - | 3 | 243 | |||
| BC025563.1 | - | 6 | 887 | 5322893 | ||
| BC065167.1 | - | 5 | 588 | |||
| CB247282.1 | - | 3 | 239 | |||
| CK619719.1 | - | 4 | 497 | |||

Klhl18
Genomic Map