siRNA Details
| Gene Symbol: Armc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177778.4 | NP_808446.1 | 1, 2 | 348 | ||
| GenBank mRNA | AK042345.1 | - | 1, 2 | 184 | ||
| AK146249.1 | - | 1, 2 | 310 | |||
| BC023126.1 | - | 1, 2 | 348 | 2616092 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Armc7| Gene Symbol: Armc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177778.4 | NP_808446.1 | 1, 2 | 348 | ||
| GenBank mRNA | AK042345.1 | - | 1, 2 | 184 | ||
| AK146249.1 | - | 1, 2 | 310 | |||
| BC023126.1 | - | 1, 2 | 348 | 2616092 | ||