siRNA Details
| Gene Symbol: Hist1h2ae | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK028026.1 | - | 1 | 560 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hist1h2ae| Gene Symbol: Hist1h2ae | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK028026.1 | - | 1 | 560 | ||