siRNA Details
| Gene Symbol: Tmem30b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178715.3 | NP_848830.1 | 1 | 1534 | ||
| GenBank mRNA | AK030302.1 | - | 1 | 1529 | ||
| AK033580.1 | - | 1 | 1535 | |||
| AK037246.1 | - | 1 | 1532 | |||
| AK169014.1 | - | 1 | 1570 | |||
| BC119224.1 | - | 1 | 1527 | 8733973 | ||
| BC119226.1 | - | 1 | 1527 | 8733975 | ||

Tmem30b
Genomic Map