siRNA Details
| Gene Symbol: Mief1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178719.5 | NP_848834.2 | 6 | 3347 | ||
| GenBank mRNA | AK041184.1 | - | 6 | 3240 | ||
| AK050546.1 | - | 6 | 3307 | |||
| AK080375.1 | - | 6 | 3297 | |||
| AK163389.1 | - | 8 | 2383 | |||
| BC094597.1 | - | 6 | 3279 | |||

Mief1
Genomic Map