siRNA Details
| Gene Symbol: Zpld1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178720.4 | NP_848835.1 | 6, 7 | 749 | ||
| GenBank mRNA | AK034628.1 | - | 6, 7 | 750 | ||
| AK047981.1 | - | 5, 6 | 659 | |||
| AK052599.1 | - | 7, 8 | 859 | |||
| BC116254.1 | - | 6, 7 | 718 | 40093631 | ||
| BC116255.1 | - | 6, 7 | 718 | 40093632 | ||

Zpld1
Genomic Map