siRNA Details
| Gene Symbol: Zpld1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178720.4 | NP_848835.1 | 12 | 2108 | ||
| GenBank mRNA | AK034628.1 | - | 12 | 2109 | ||
| AK047981.1 | - | 11 | 2016 | |||
| BC116254.1 | - | 12 | 2077 | 40093631 | ||
| BC116255.1 | - | 12 | 2077 | 40093632 | ||

Zpld1
Genomic Map