siRNA Details
| Gene Symbol: Ppm1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178726.3 | NP_848841.2 | 2 | 1063 | ||
| GenBank mRNA | AK028275.1 | - | 2 | 105 | ||
| AK032529.1 | - | 2 | 1121 | |||
| AK035912.1 | - | 2 | 1066 | |||
| AK045724.1 | - | 2 | 1104 | |||
| AK131646.1 | - | 2 | 1050 | |||
| AK142789.1 | - | 2 | 1081 | |||
| AK147389.1 | - | 2 | 1066 | |||
| AK147876.1 | - | 2 | 1100 | |||
| AK165160.1 | - | 2 | 1096 | |||
| AK220523.1 | - | 2 | 1068 | |||
| BC096031.1 | - | 2 | 1063 | 6837066 | ||

Ppm1l
Genomic Map