siRNA Details
| Gene Symbol: D630039A03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178727.2 | NP_848842.1 | 1 | 583 | ||
| GenBank mRNA | AK033446.1 | - | 1 | 87 | ||
| AK035369.1 | - | 1 | 617 | |||
| AK085537.1 | - | 1 | 564 | |||
| BC030929.1 | - | 1 | 535 | 4911378 | ||
| BC095953.1 | - | 1 | 583 | 5043405 | ||

D630039A03Rik
Genomic Map