siRNA Details
| Gene Symbol: Zfp324 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178732.3 | NP_848847.3 | 4 | 1284 | ||
| GenBank mRNA | AK044240.1 | - | 4 | 1281 | ||
| AK051488.1 | - | 4 | 1307 | |||
| AK052251.1 | - | 3 | 1177 | |||
| AK054407.1 | - | 4 | 1329 | |||
| AK085053.1 | - | 3 | 1180 | |||
| AK152107.1 | - | 4 | 1310 | |||
| AK152696.1 | - | 4 | 1227 | |||
| AK165922.1 | - | 4 | 1280 | |||
| BC053093.1 | - | 4 | 1284 | 6400158 | ||

Zfp324
Genomic Map