siRNA Details
| Gene Symbol: Elmod2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170691.1 | NP_001164162.1 | 2 | 242 | ||
| NM_178736.5 | NP_848851.3 | 2 | 247 | |||
| GenBank mRNA | AK029044.1 | - | 2 | 218 | ||
| AK036607.1 | - | 2 | 225 | |||
| AK036746.1 | - | 2 | 187 | |||
| AK041137.1 | - | 2 | 148 | |||
| AK082730.1 | - | 1 | 14 | |||
| AK084950.1 | - | 2 | 247 | |||
| AK165222.1 | - | 2 | 166 | |||
| BC079654.1 | - | 2 | 208 | 6855187 | ||

Elmod2
Genomic Map