siRNA Details
| Gene Symbol: Dcaf12l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190718.1 | NP_001177647.1 | 1 | 1076 | ||
| NM_178739.6 | NP_848854.3 | 1 | 1112 | |||
| GenBank mRNA | AK034343.1 | - | 1 | 929 | ||
| AK034472.1 | - | 1 | 1112 | |||
| AK038557.1 | - | 1 | 1067 | |||
| AK039767.1 | - | 1 | 933 | |||
| AK047360.1 | - | 1 | 1076 | |||
| BC068319.1 | - | 1 | 916 | 30431243 | ||

Dcaf12l1
Genomic Map