siRNA Details
| Gene Symbol: Klhl8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178741.3 | NP_848856.1 | 4 | 1155 | ||
| GenBank mRNA | AK031028.1 | - | 4 | 1156 | ||
| AK034618.1 | - | 4 | 1121 | |||
| AK049960.1 | - | 3 | 1043 | |||
| AK209067.1 | - | 2 | 149 | |||
| AK211422.1 | - | 2 | 150 | |||
| AY101769.1 | - | 3 | 900 | |||
| BC086802.1 | - | 3 | 1016 | 30612176 | ||

Klhl8
Genomic Map