siRNA Details
| Gene Symbol: Slc38a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178746.4 | NP_848861.1 | 3 | 345 | ||
| GenBank mRNA | AK043402.1 | - | 3 | 333 | ||
| AK044735.1 | - | 3 | 335 | |||
| BC052361.1 | - | 3 | 345 | 6705789 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc38a9| Gene Symbol: Slc38a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178746.4 | NP_848861.1 | 3 | 345 | ||
| GenBank mRNA | AK043402.1 | - | 3 | 333 | ||
| AK044735.1 | - | 3 | 335 | |||
| BC052361.1 | - | 3 | 345 | 6705789 | ||