siRNA Details
| Gene Symbol: Spin4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178753.4 | NP_848868.1 | 1 | 1021 | ||
| GenBank mRNA | AK033181.1 | - | 1 | 338 | ||
| AK036158.1 | - | 1 | 1022 | |||
| AK043030.1 | - | 1 | 989 | |||
| AK047896.1 | - | 1 | 982 | |||
| AK147852.1 | - | 1 | 988 | |||
| AK155170.1 | - | 1 | 961 | |||
| BC027796.1 | - | 1 | 987 | 5323183 | ||

Spin4
Genomic Map