siRNA Details
| Gene Symbol: Lppr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178756.4 | NP_848871.1 | 3 | 328 | ||
| GenBank mRNA | AK034251.1 | - | 3 | 513 | ||
| AK043762.1 | - | 3 | 230 | |||
| AK047614.1 | - | 3 | 258 | |||
| AK053451.1 | - | 3 | 334 | |||
| AK087439.1 | - | 3 | 479 | |||
| AK087514.1 | - | 3 | 329 | |||
| AK160897.1 | - | 3 | 455 | |||
| AY345342.1 | - | 3 | 328 | |||
| BC096762.1 | - | 3 | 304 | 30094664 | ||

Lppr1
Genomic Map