siRNA Details
| Gene Symbol: Zfp750 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178763.4 | NP_848878.1 | 3 | 3096 | ||
| GenBank mRNA | AK037185.1 | - | 3 | 3057 | ||
| AK140792.1 | - | 3091 | ||||
| CF586141.1 | - | 1 | 104 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp750| Gene Symbol: Zfp750 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178763.4 | NP_848878.1 | 3 | 3096 | ||
| GenBank mRNA | AK037185.1 | - | 3 | 3057 | ||
| AK140792.1 | - | 3091 | ||||
| CF586141.1 | - | 1 | 104 | |||