siRNA Details
| Gene Symbol: Tmem72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178768.4 | NP_848883.2 | 5 | 1417 | ||
| GenBank mRNA | AK033856.1 | - | 5 | 1554 | ||
| AK049048.1 | - | 5 | 1572 | |||
| AK049938.1 | - | 5 | 1530 | |||
| AK085603.1 | - | 5 | 1418 | |||
| AK141578.1 | - | 5 | 1574 | |||
| BC093486.1 | - | 5 | 1560 | 30697044 | ||

Tmem72
Genomic Map