siRNA Details
| Gene Symbol: Aoc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178932.1 | NP_849263.1 | 4 | 2376 | ||
| GenBank mRNA | AF350445.2 | - | 4 | 2376 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aoc2| Gene Symbol: Aoc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178932.1 | NP_849263.1 | 4 | 2376 | ||
| GenBank mRNA | AF350445.2 | - | 4 | 2376 | ||