siRNA Details
| Gene Symbol: Aoc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178932.1 | NP_849263.1 | 1, 2 | 1616 | ||
| GenBank mRNA | AF350445.2 | - | 1, 2 | 1616 | ||
| BC150843.1 | - | 1, 2 | 1607 | 9087754 | ||
| BC150848.1 | - | 1, 2 | 1607 | 9087759 | ||

Aoc2
Genomic Map